Assay Details
Gene Symbol: LOC5574609 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845802.1 | XP_021701494.1 | 5 - 6 | 3362 | 68 | |
XM_021845803.1 | XP_021701495.1 | 3 - 4 | 2124 | 68 | ||
XM_021845804.1 | XP_021701496.1 | 3 - 4 | 2845 | 68 |

More Information
Set Membership: |
![]() ![]() |
