Assay Details
Gene Symbol: LOC5564337 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021851565.1 | XP_021707257.1 | 4 - 5 | 682 | 87 | |
XM_021851566.1 | XP_021707258.1 | 3 - 4 | 1038 | 87 |
