Assay Details
Gene Symbol: LOC5564337 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021851565.1 | XP_021707257.1 | 12 - 13 | 1703 | 73 | |
XM_021851566.1 | XP_021707258.1 | 11 - 12 | 2059 | 73 |
