Assay Details
Gene Symbol: LOC5564337 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021851565.1 | XP_021707257.1 | 11 - 12 | 1546 | 100 | |
XM_021851566.1 | XP_021707258.1 | 10 - 11 | 1902 | 100 |
