Assay Details
Gene Symbol: LOC5565494 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001649790.2 | XP_001649840.1 | 1 - 2 | 235 | 68 | |
XM_001649791.2 | XP_001649841.1 | 1 - 2 | 236 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
