Assay Details
Gene Symbol: LOC5572049 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001660105.2 | XP_001660155.1 | 2 - 3 | 321 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC5572049 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001660105.2 | XP_001660155.1 | 2 - 3 | 321 | 99 |
Set Membership: |
![]() ![]() ![]() ![]() |