Assay Details
Gene Symbol: LOC5569462 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021855198.1 | XP_021710890.1 | 3 - 4 | 1580 | 83 | |
XM_021855207.1 | XP_021710899.1 | 3 - 4 | 1321 | 83 | ||
XM_021855213.1 | XP_021710905.1 | 4 - 5 | 2063 | 83 | ||
XM_021855223.1 | XP_021710915.1 | 5 - 6 | 1791 | 83 | ||
XM_021855229.1 | XP_021710921.1 | 5 - 6 | 1707 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |
