Assay Details
Gene Symbol: LOC5569462 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021855213.1 | XP_021710905.1 | 2 - 3 | 1089 | 114 | |
XM_021855223.1 | XP_021710915.1 | 3 - 4 | 817 | 114 | ||
XM_021855229.1 | XP_021710921.1 | 3 - 4 | 733 | 114 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021855213.1, XM_021855223.1, XM_021855229.1 transcripts.
Set Membership: |
![]() ![]() |
