Assay Details
Gene Symbol: LOC5564986 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845663.1 | XP_021701355.1 | 5 - 6 | 2128 | 84 | |
XM_021845664.1 | XP_021701356.1 | 5 - 6 | 2109 | 84 | ||
XM_021845665.1 | XP_021701357.1 | 5 - 6 | 2106 | 84 | ||
XM_021845666.1 | XP_021701358.1 | 5 - 6 | 1897 | 84 | ||
XM_021845667.1 | XP_021701359.1 | 6 - 7 | 2400 | 84 | ||
XM_021845668.1 | XP_021701360.1 | 5 - 6 | 2105 | 84 | ||
XM_021845669.1 | XP_021701361.1 | 5 - 6 | 2091 | 84 | ||
XM_021845671.1 | XP_021701363.1 | 5 - 6 | 2050 | 84 | ||
XM_021845672.1 | XP_021701364.1 | 4 - 5 | 1441 | 84 | ||
XM_021845673.1 | XP_021701365.1 | 5 - 6 | 2274 | 84 | ||
XM_021845674.1 | XP_021701366.1 | 7 - 8 | 1940 | 84 | ||
XM_021845675.1 | XP_021701367.1 | 8 - 9 | 2105 | 84 | ||
XM_021845676.1 | XP_021701368.1 | 8 - 9 | 2122 | 84 |
