Assay Details
Gene Symbol: LOC5578894 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001664052.2 | XP_001664102.2 | 3 - 4 | 926 | 81 | |
XM_021844439.1 | XP_021700131.1 | 3 - 4 | 929 | 81 | ||
XM_021844440.1 | XP_021700132.1 | 3 - 4 | 881 | 81 | ||
XM_021844441.1 | XP_021700133.1 | 3 - 4 | 903 | 81 | ||
XM_021844442.1 | XP_021700134.1 | 3 - 4 | 900 | 81 | ||
XM_021844443.1 | XP_021700135.1 | 3 - 4 | 874 | 81 |
