Assay Details
Gene Symbol: LOC5578188 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021851237.1 | XP_021706929.1 | 1 - 2 | 588 | 116 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021851237.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
