Assay Details
Gene Symbol: LOC5567813 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021839879.1 | XP_021695571.1 | 12 - 13 | 4467 | 73 | |
XM_021839880.1 | XP_021695572.1 | 12 - 13 | 4387 | 73 | ||
XM_021839882.1 | XP_021695574.1 | 11 - 12 | 4692 | 73 | ||
XM_021839883.1 | XP_021695575.1 | 10 - 11 | 3264 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
