Assay Details
Gene Symbol: LOC5567813 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021839879.1 | XP_021695571.1 | 2 - 3 | 736 | 58 | |
XM_021839880.1 | XP_021695572.1 | 2 - 3 | 656 | 58 | ||
XM_021839882.1 | XP_021695574.1 | 1 - 2 | 961 | 58 | ||
XM_021839883.1 | XP_021695575.1 | 1 - 2 | 952 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() |
