Assay Details
Gene Symbol: LOC5567813 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021839879.1 | XP_021695571.1 | 10 - 11 | 3765 | 69 | |
XM_021839880.1 | XP_021695572.1 | 10 - 11 | 3685 | 69 | ||
XM_021839882.1 | XP_021695574.1 | 9 - 10 | 3990 | 69 | ||
XM_021839883.1 | XP_021695575.1 | 8 - 9 | 2562 | 69 |

More Information
Set Membership: |
![]() ![]() |
