Assay Details
Gene Symbol: LOC5568890 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001658166.2 | XP_001658216.2 | 3 - 4 | 1424 | 75 | |
XM_021845772.1 | XP_021701464.1 | 3 - 4 | 1428 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
