Assay Details
Gene Symbol: LOC5578213 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001656807.2 | XP_001656857.2 | 6 - 7 | 1710 | 63 | |
XM_021839948.1 | XP_021695640.1 | 6 - 7 | 1829 | 63 | ||
XM_021839950.1 | XP_021695642.1 | 6 - 7 | 1578 | 63 | ||
XM_021839951.1 | XP_021695643.1 | 6 - 7 | 1540 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
