Assay Details
Gene Symbol: LOC5578213 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001656807.2 | XP_001656857.2 | 7 - 8 | 1960 | 63 | |
XM_021839948.1 | XP_021695640.1 | 7 - 8 | 2079 | 63 | ||
XM_021839950.1 | XP_021695642.1 | 7 - 8 | 1828 | 63 | ||
XM_021839951.1 | XP_021695643.1 | 7 - 8 | 1790 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
