Assay Details
Gene Symbol: LOC5578213 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001656807.2 | XP_001656857.2 | 8 - 9 | 2047 | 63 | |
XM_021839948.1 | XP_021695640.1 | 8 - 9 | 2166 | 63 | ||
XM_021839950.1 | XP_021695642.1 | 8 - 9 | 1915 | 63 | ||
XM_021839951.1 | XP_021695643.1 | 8 - 9 | 1877 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
