Assay Details
Gene Symbol: LOC5576045 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001662320.2 | XP_001662370.2 | 4 - 5 | 980 | 97 | |
XM_001662321.2 | XP_001662371.2 | 3 - 4 | 1056 | 97 | ||
XM_021842617.1 | XP_021698309.1 | 3 - 4 | 952 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() |
