Assay Details
Gene Symbol: LOC5563742 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001647980.2 | XP_001648030.2 | 1 - 2 | 395 | 73 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001647980.2 transcripts.
Set Membership: |
![]() ![]() |
