Assay Details
Gene Symbol: LOC5569795 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021853677.1 | XP_021709369.1 | 4 - 5 | 1577 | 56 | |
XM_021853678.1 | XP_021709370.1 | 4 - 5 | 1917 | 56 | ||
XM_021853679.1 | XP_021709371.1 | 4 - 5 | 1561 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
