Assay Details
Gene Symbol: LOC5566237 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021841148.1 | XP_021696840.1 | 6 - 7 | 2165 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC5566237 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021841148.1 | XP_021696840.1 | 6 - 7 | 2165 | 75 |
Set Membership: |
![]() ![]() ![]() |