Assay Details
Gene Symbol: LOC5566237 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001650562.2 | XP_001650612.2 | 1 - 2 | 929 | 61 | |
XM_021841148.1 | XP_021696840.1 | 1 - 2 | 930 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
