Assay Details
Gene Symbol: LOC5575098 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021853136.1 | XP_021708828.1 | 4 - 5 | 2480 | 107 | |
XM_021853137.1 | XP_021708829.1 | 4 - 5 | 3572 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() |
