Assay Details
Gene Symbol: LOC5575098 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021853136.1 | XP_021708828.1 | 2 - 3 | 1394 | 92 | |
XM_021853137.1 | XP_021708829.1 | 2 - 3 | 2486 | 92 |
