Assay Details
Gene Symbol: LOC5570012 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021854804.1 | XP_021710496.1 | 8 - 9 | 2995 | 81 | |
XM_021854805.1 | XP_021710497.1 | 8 - 9 | 3045 | 81 | ||
XM_021854806.1 | XP_021710498.1 | 6 - 7 | 2467 | 81 | ||
XM_021854807.1 | XP_021710499.1 | 6 - 7 | 2092 | 81 |
