Assay Details
Gene Symbol: LOC5570012 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021854804.1 | XP_021710496.1 | 7 - 8 | 2576 | 73 | |
XM_021854805.1 | XP_021710497.1 | 7 - 8 | 2626 | 73 | ||
XM_021854806.1 | XP_021710498.1 | 5 - 6 | 2048 | 73 | ||
XM_021854807.1 | XP_021710499.1 | 5 - 6 | 1673 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
