Assay Details
Gene Symbol: LOC5572215 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021853022.1 | XP_021708714.1 | 1 - 2 | 1415 | 87 | |
XM_021853027.1 | XP_021708719.1 | 1 - 2 | 1412 | 87 | ||
XM_021853028.1 | XP_021708720.1 | 1 - 2 | 1422 | 87 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021853022.1, XM_021853027.1, XM_021853028.1 transcripts.
Set Membership: |
![]() ![]() |
