Assay Details
Gene Symbol: LOC5572215 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021853022.1 | XP_021708714.1 | 8 - 9 | 4724 | 68 | |
XM_021853023.1 | XP_021708715.1 | 6 - 7 | 4208 | 68 | ||
XM_021853024.1 | XP_021708716.1 | 6 - 7 | 4156 | 68 | ||
XM_021853025.1 | XP_021708717.1 | 3 - 4 | 1974 | 68 | ||
XM_021853027.1 | XP_021708719.1 | 8 - 9 | 4721 | 68 | ||
XM_021853028.1 | XP_021708720.1 | 8 - 9 | 4731 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
