Assay Details
Gene Symbol: LOC5572215 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021853022.1 | XP_021708714.1 | 7 - 8 | 4399 | 79 | |
XM_021853023.1 | XP_021708715.1 | 5 - 6 | 3883 | 79 | ||
XM_021853024.1 | XP_021708716.1 | 5 - 6 | 3831 | 79 | ||
XM_021853025.1 | XP_021708717.1 | 2 - 3 | 1649 | 79 | ||
XM_021853027.1 | XP_021708719.1 | 7 - 8 | 4396 | 79 | ||
XM_021853028.1 | XP_021708720.1 | 7 - 8 | 4406 | 79 |
