Assay Details
Gene Symbol: LOC5575522 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021848028.1 | XP_021703720.1 | 4 - 5 | 1015 | 72 | |
XM_021848029.1 | XP_021703721.1 | 3 - 4 | 774 | 72 | ||
XM_021848030.1 | XP_021703722.1 | 3 - 4 | 761 | 72 | ||
XM_021848031.1 | XP_021703723.1 | 3 - 4 | 764 | 72 | ||
XM_021848032.1 | XP_021703724.1 | 3 - 4 | 749 | 72 | ||
XM_021848033.1 | XP_021703725.1 | 3 - 4 | 719 | 72 | ||
XM_021848034.1 | XP_021703726.1 | 4 - 5 | 795 | 72 | ||
XM_021848035.1 | XP_021703727.1 | 3 - 4 | 627 | 72 | ||
XM_021848036.1 | XP_021703728.1 | 4 - 5 | 703 | 72 | ||
XM_021848037.1 | XP_021703729.1 | 3 - 4 | 630 | 72 | ||
XM_021848039.1 | XP_021703731.1 | 2 - 3 | 447 | 72 | ||
XM_021848040.1 | XP_021703732.1 | 3 - 4 | 350 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
