Assay Details
Gene Symbol: LOC5568734 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021842325.1 | XP_021698017.1 | 10 - 11 | 2857 | 63 | |
XM_021842326.1 | XP_021698018.1 | 10 - 11 | 2962 | 63 | ||
XM_021842327.1 | XP_021698019.1 | 10 - 11 | 2830 | 63 | ||
XM_021842329.1 | XP_021698021.1 | 10 - 11 | 2809 | 63 | ||
XM_021842330.1 | XP_021698022.1 | 10 - 11 | 2914 | 63 | ||
XM_021842331.1 | XP_021698023.1 | 9 - 10 | 2524 | 63 | ||
XM_021842332.1 | XP_021698024.1 | 9 - 10 | 2628 | 63 | ||
XM_021842333.1 | XP_021698025.1 | 9 - 10 | 2602 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
