Assay Details
Gene Symbol: LOC5568734 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021842326.1 | XP_021698018.1 | 9 - 10 | 2779 | 87 | |
XM_021842330.1 | XP_021698022.1 | 9 - 10 | 2731 | 87 | ||
XM_021842332.1 | XP_021698024.1 | 8 - 9 | 2445 | 87 | ||
XM_021842333.1 | XP_021698025.1 | 8 - 9 | 2419 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
