Assay Details
Gene Symbol: LOC5568734 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021842325.1 | XP_021698017.1 | 8 - 9 | 2496 | 82 | |
XM_021842326.1 | XP_021698018.1 | 8 - 9 | 2613 | 82 | ||
XM_021842327.1 | XP_021698019.1 | 8 - 9 | 2469 | 82 | ||
XM_021842329.1 | XP_021698021.1 | 8 - 9 | 2448 | 82 | ||
XM_021842330.1 | XP_021698022.1 | 8 - 9 | 2565 | 82 | ||
XM_021842331.1 | XP_021698023.1 | 7 - 8 | 2163 | 82 | ||
XM_021842332.1 | XP_021698024.1 | 7 - 8 | 2279 | 82 | ||
XM_021842333.1 | XP_021698025.1 | 7 - 8 | 2253 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
