Assay Details
Gene Symbol: LOC5568734 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021842325.1 | XP_021698017.1 | 7 - 8 | 2119 | 65 | |
XM_021842326.1 | XP_021698018.1 | 7 - 8 | 2236 | 65 | ||
XM_021842327.1 | XP_021698019.1 | 7 - 8 | 2092 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
