Assay Details
Gene Symbol: LOC5573478 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001654575.2 | XP_001654625.2 | 3 - 4 | 1370 | 100 | |
XM_021840323.1 | XP_021696015.1 | 3 - 4 | 1364 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
