Assay Details
Gene Symbol: LOC5579439 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021846872.1 | XP_021702564.1 | 5 - 6 | 3390 | 75 | |
XM_021846880.1 | XP_021702572.1 | 3 - 4 | 1115 | 75 | ||
XM_021846885.1 | XP_021702577.1 | 3 - 4 | 1100 | 75 | ||
XM_021846893.1 | XP_021702585.1 | 2 - 3 | 606 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
