Assay Details
Gene Symbol: LOC5564565 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021837839.1 | XP_021693531.1 | 12 - 13 | 5292 | 80 | |
XM_021837840.1 | XP_021693532.1 | 12 - 13 | 5360 | 80 | ||
XM_021837841.1 | XP_021693533.1 | 12 - 13 | 5295 | 80 | ||
XM_021837842.1 | XP_021693534.1 | 13 - 14 | 5161 | 80 | ||
XM_021837843.1 | XP_021693535.1 | 12 - 13 | 4983 | 80 | ||
XM_021837844.1 | XP_021693536.1 | 13 - 14 | 3410 | 80 | ||
XM_021837845.1 | XP_021693537.1 | 12 - 13 | 3383 | 80 | ||
XM_021837846.1 | XP_021693538.1 | 11 - 12 | 3121 | 80 | ||
XM_021837847.1 | XP_021693539.1 | 11 - 12 | 3124 | 80 | ||
XM_021837848.1 | XP_021693540.1 | 5 - 6 | 1219 | 80 | ||
XM_021837850.1 | XP_021693542.1 | 4 - 5 | 1192 | 80 |
