Assay Details
Gene Symbol: LOC5567075 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001651421.2 | XP_001651471.2 | 9 - 10 | 1222 | 64 | |
XM_021857224.1 | XP_021712916.1 | 9 - 10 | 1229 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
