Assay Details
Gene Symbol: LOC5575094 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001661768.2 | XP_001661818.2 | 1 - 2 | 538 | 110 | |
XM_021850281.1 | XP_021705973.1 | 1 - 2 | 539 | 110 |

More Information
Set Membership: |
![]() ![]() |
