Assay Details
Gene Symbol: LOC5574617 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021857661.1 | XP_021713353.1 | 2 - 3 | 819 | 105 | |
XM_021857662.1 | XP_021713354.1 | 2 - 3 | 819 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() |
