Assay Details
Gene Symbol: LOC5574617 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021857661.1 | XP_021713353.1 | 8 - 9 | 3686 | 56 | |
XM_021857662.1 | XP_021713354.1 | 8 - 9 | 3686 | 56 | ||
XM_021857663.1 | XP_021713355.1 | 7 - 8 | 3611 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
