Assay Details
Gene Symbol: LOC5565618 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021844844.1 | XP_021700536.1 | 12 - 13 | 3878 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC5565618 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021844844.1 | XP_021700536.1 | 12 - 13 | 3878 | 68 |
Set Membership: |
![]() ![]() ![]() ![]() |