Assay Details
Gene Symbol: LOC5565618 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021844844.1 | XP_021700536.1 | 11 - 12 | 3722 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC5565618 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021844844.1 | XP_021700536.1 | 11 - 12 | 3722 | 81 |
Set Membership: |
![]() ![]() ![]() |