Assay Details
Gene Symbol: LOC5579092 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021850433.1 | XP_021706125.1 | 15 - 16 | 3445 | 98 | |
XM_021850434.1 | XP_021706126.1 | 14 - 15 | 3544 | 98 | ||
XM_021850435.1 | XP_021706127.1 | 14 - 15 | 3527 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
