Assay Details
Gene Symbol: LOC5579092 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021850433.1 | XP_021706125.1 | 2 - 3 | 1175 | 79 | |
XM_021850434.1 | XP_021706126.1 | 1 - 2 | 1274 | 79 | ||
XM_021850435.1 | XP_021706127.1 | 1 - 2 | 1275 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
