Assay Details
Gene Symbol: LOC5579092 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021850433.1 | XP_021706125.1 | 8 - 9 | 1926 | 68 | |
XM_021850434.1 | XP_021706126.1 | 7 - 8 | 2025 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
