Assay Details
Gene Symbol: LOC5579092 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021850433.1 | XP_021706125.1 | 9 - 10 | 2146 | 65 | |
XM_021850434.1 | XP_021706126.1 | 8 - 9 | 2245 | 65 | ||
XM_021850435.1 | XP_021706127.1 | 8 - 9 | 2228 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
