Assay Details
Gene Symbol: LOC5579092 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021850433.1 | XP_021706125.1 | 10 - 11 | 2292 | 76 | |
XM_021850434.1 | XP_021706126.1 | 9 - 10 | 2391 | 76 | ||
XM_021850435.1 | XP_021706127.1 | 9 - 10 | 2374 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
