Assay Details
Gene Symbol: LOC5577130 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021856919.1 | XP_021712611.1 | 6 - 7 | 1559 | 103 | |
XM_021856923.1 | XP_021712615.1 | 5 - 6 | 1655 | 103 |

More Information
Set Membership: |
![]() ![]() |
